ClipTip® 384 Pipette Tips, 30 μl, Thermo Scientific
Supplier: Thermo Scientific
94410100 94410103 94420103
10030-802CS 1329.58 CAD
10030-802 10030-804 10030-806
ClipTip® 384 Pipette Tips, 30 μl, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® 384 pipette tips feature a unique ‘Snap and Lip’ seal technology to attach and seal the tips on each channel with minimal tip attachment and ejection force.
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Each tip insert has ink-jetted lot and volume information
ClipTip® 384 model selection is optimized for 384-microplate and molecular biology applications. The tips are available in three sizes and packaging options, with 12.5, 30, and 125 μl tips available in nonsterile, sterile, and filtered formats. AMore Product Information