Advanced Purpose Heating Circulators, Grant Instruments
Supplier: GRANT USA INC.
TXF200 US TX150 EURO TX150-ST5 US TX150-ST12 US TX150-ST18 US TX150-ST26 US TX150-ST38 US TXF200-ST5 US TXF200-ST12 US TXF200-ST18 US TXF200-ST26 US
89260-844EA 8473.73 CAD
89260-844 89260-850 10029-470 10029-472 10029-474 10029-476 10029-478 10029-480 10029-482 10029-484 10029-488 10029-490 10029-492 10029-494 10029-496 10029-498 10029-506 10029-508
Advanced Purpose Heating Circulators, Grant Instruments
Circulating Thermostats Closed Circulator Systems Heating Circulators
The Optima™ range of advanced thermostats can be combined with plastic baths (5L, 12L, 18L), premium stainless steel baths (5L, 12L, 18L, 26L, 38L) or refrigeration units (3 units available)
Additional features include countdown timer, adjustable over-temperature cut-out, visual, audible and programmable high and low temperature alarms and three temperature presets. The TX150 has the capacity to store one programme with 30 segments. The TXF200 can also be programmed via the user interface and can store up to 10 programmes containing 100 segments. The programming interface includes setMore Product Information