Natural Air Circulation Incubators Units, Memmert
Supplier: Memmert
IN30, 115 V IN30, 230 V IN30PLUS115 IN30PLUS230 IN55, 115 V IN55, 230V IN55PLUS115 IN55PLUS230 IN75, 115 V IN75, 230 V IN75PLUS115 IN75PLUS230 IN110, 115 V IN110, 230 V IN110PLUS115 IN110PLUS230 IN160, 115 V IN160, 230 V IN160PLUS115 IN160PLUS230 IN260, 115 V IN260, 230 V IN260PLUS115 IN260PLUS230 IN450PLUS115 IN450PLUS230 IN750PLUS115 IN750PLUS230 IN450115V IN450230V IN750115V IN750230V
10028-930EA 3488.25 CAD
10028-930 10028-932 10028-934 10028-936 10028-938 10028-940 10028-942 10028-944 10028-946 10028-948 10028-950 10028-952 10028-954 10028-956 10028-958 10028-960 10028-962 10028-964 10028-966 10028-968 10028-970 10028-972 10028-974 10028-976 10861-392 10861-394 10861-400 10861-402 10861-388 10861-390 10861-396 10861-398
Natural Air Circulation Incubators Units, Memmert
Incubators Standard Incubators
The Generation 2012 range has natural and forced convection models with a temperature range up to +80 °C
SingleDISPLAY models; ideal for standard applications. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, programme time, time zones and daylight-saving time. One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log. Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiterMore Product Information