VWR® Microplate Shaker
Supplier: VWR International
10027-120
10027-120EA 5175.21 CAD
10027-120
VWR® Microplate Shaker
Shakers Microplate Shakers
Ideal for immunoassays and general microplate shaking, microplate shakers feature a four-place platform that holds either two or four microplates or deep well blocks.
  • Optimized for shaking microplates
  • Digital display for speed and time
  • Microprocessor control
  • Timer with audible alarm
  • Holds 2 or 4 microplates
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Plates are firmly locked into place with retaining springs. Microprocessor variable speed controlMore Product Information