VWR® Standard Line Sterile Metal-Free Centrifuge Tubes, PP
Supplier: VWR International
10025-706 10025-708 10025-710 10026-322
10025-706CS 512.35 CAD
10025-706 10025-708 10025-710 10026-322
VWR® Standard Line Sterile Metal-Free Centrifuge Tubes, PP
Tubes Centrifuge Tubes
Metal-free centrifuge tubes are ideal for environmental uses such as water analysis and other applications wherein tubes have the potential to contaminate samples.
  • Available in two volumes: 15 ml and 50 ml
  • Tubes are certified free of major critical trace metals to less than 1 ppb
  • Easy-to-read black graduations are accurate to ±2%, with 0.5 ml increments (15 ml) or 2.5 ml increments (50 ml)
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube
  • Max. rotational speed up to 12500 g
  • Autoclavable
More Product Information