VWR® Standard Line Sterile Racks for 15 and 50 ml Conical-Bottom Tubes, PP
Supplier: VWR International
10025-704 10026-064 10026-066 10026-068 10026-070 10026-300 10026-302 10026-304 10026-306 10026-308 10026-310 10026-312 10026-314 10026-316 10026-318 10026-320
10025-704CS 563.23 CAD
10025-704 10026-064 10026-066 10026-068 10026-070 10026-300 10026-302 10026-304 10026-306 10026-308 10026-310 10026-312 10026-314 10026-316 10026-318 10026-320
VWR® Standard Line Sterile Racks for 15 and 50 ml Conical-Bottom Tubes, PP
Racks Tube Racks
Polypropylene racks are available in seven colors, and are also offered in an assorted color pack with one of each color: blue, green, orange, red, and yellow.
  • Available in a variety of colors
  • Suitable for plug or flat cap tubes
  • Polypropylene construction
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Racks accommodate plug or flat cap tubes.
Certifications: TSE/BSE-free Certification.
Ordering information: All racks are sterilized by irradiation SAL10-6,More Product Information