Large-Digit Triple Display Thermometers, Thermco
Supplier: Thermco
ACC895FREV ACC895INCV ACC895REFV
10025-458EA 191.04 CAD
10025-458 10025-460 10025-462
Large-Digit Triple Display Thermometers, Thermco
Thermometers Digital Thermometers
Triple display digital thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously.
  • NIST Compliant®
  • Each thermometer is individually serialized
  • °C and °F switchable
The Triple Display Digital Thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously.
An audible alarm will sound when the temperature rises above or goes below the set temperatures. Thermometer probe is housed in an unbreakable plastic bottle or vial filled with a Bio-Safe glycol/water or sandMore Product Information