Digital Stopwatch, Industrial Physics
Supplier: GREAT LAKES GAGES, LLC
DI0076
10025-184EA 167.04 CAD
10025-184
Digital Stopwatch, Industrial Physics
Timers Digital Stopwatches
Accurate digital TQC stopwatch, resolution to ¹/₁₀₀th of a second.
  • Split times, built-in alarm
  • Clear display with large digits
  • Indispensible for viscosity measurements
Ultra flat and robust water- and shock proof design, complete with carrying cord.