MedTest 200 Ensemble, MedTest
Supplier: MEDTEST DX, INC.
BS200/ISE
10023-854EA 130078.35 CAD
10023-854
MedTest 200 Ensemble, MedTest
Analysers Elemental Analysers
The MedTest 200 Ensemble provides exceptional value and convenience by bringing together low to mid volume laboratory needs to perform clinical chemistry testing.
  • Light source provided by halogen-tungsten lamp
  • 200 tests per hour/up to 330 tests per hour with ISE
  • Bi-directional LIS interface transmission
  • Reversed optic system with 8 wavelengths from 340 to 670 nm
Discrete, random access and automated. Automatic probe cleaning, liquid level detection, horizontal and vertical collision protection.