VWR® Basic Reach-In Freezers with Natural Refrigerant
Supplier: VWR International
10018-870 75846-302 75876-102
10018-870EA 16098.73 CAD
10018-870 75846-302 75876-102
VWR® Basic Reach-In Freezers with Natural Refrigerant
Freezers Laboratory / General Purpose Freezers
Upright freezers with 1-, 2-, or 3-doors, these freezers maintain –10 °F (–23.3 °C).
  • Environmentally friendly
  • Stainless steel solid door and front, aluminium sides, with white interior
  • Swinging doors
  • Automatic defrost
  • Lifetime guaranteed recessed door handles and door hinges
Standard with 3, 6, or 9 adjustable heavy duty PVC coated full gray shelves and LED lighting.
HC - Natural refrigerant that is environmentally friendly R290 hydro carbon refrigerant that has zero (0) ozone depletion potential (ODP), and three (3) global warming potential (GWP).
More Product Information