VWR® Basic Refrigerators with Glass Doors and Natural Refrigerant
Supplier: VWR International
10791-618 10791-624 10791-626 10018-860 10018-864 10018-866 75871-648 75871-650
10791-618EA 12272.32 CAD
10791-618 10791-624 10791-626 10018-860 10018-864 10018-866 75871-648 75871-650
VWR® Basic Refrigerators with Glass Doors and Natural Refrigerant
Refrigerators
Swinging or sliding doors. These upright laboratory refrigerators maintain an average temperature of 3 – 5 °C (37 – 41 °F).
  • All units feature a natural refrigerant that is environmentally friendly
  • R290 hydrocarbon refrigerant emits zero ozone depletion potential
  • Units are supplied standard with the LAE temperature control
  • Exterior digital display and alarm function
Units are equipped with low E, double-pane, thermal insulated glass doors for energy efficiency.
Note: Units do not come with castors. Castors can be ordered separately.