Low-Halide Polyester Labels, B30, Brady
Supplier: Brady Worldwide
117958 117957 117968 117967 117940 117939 117938 117937 117952 117951 117950 117949 117956 117955 117954 117953 117960 117959 117966 117965 117972 117971 117970 117969 117980 117979 117978 117984 117983 117982 117981
10017-994EA 522.56 CAD
10017-994 10017-996 10017-998 10018-000 10018-052 10018-054 10018-056 10018-058 10018-060 10018-450 10018-452 10018-454 10018-456 10018-458 10018-460 10018-462 10018-464 10018-466 10018-468 10018-470 10018-472 10018-474 10018-476 10018-478 10018-480 10018-482 10018-484 10018-486 10018-488 10018-490 10018-492 10018-494
Low-Halide Polyester Labels, B30, Brady
Labels
Hi-Performance Polyester Labels (B-569) have a low halide content and are designed for marking stainless steel pipes, particularly in harsh environments.
  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester label can be thermal transfer printed
  • Withstands mild chemicals, oil, and water
  • Service temperature of –40 to 100 °C (–40 to 212 °F)
  • For use with BBP®30; BBP®31; BBP®33; BBP®35; BBP®37 printers
Applies best to very smooth surfaces and ideal for mounting to rigid panels. Continuous roll isMore Product Information