DuPont™ Tyvek® 400 D Coveralls, Comfort Fit Design
Supplier: DuPont
TD125SWB2X0025CM TD125SWB3X0025CM TD125SWB4X0025CM TD125SWBMD0025CM TD125SWBLG0025CM TD125SWBXL0025CM
10016-534CS 508 CAD
10016-534 10016-536 10016-538 10016-542 CA10016-540 CA10016-544
DuPont™ Tyvek® 400 D Coveralls, Comfort Fit Design
Disposable Garments
DuPont™ Tyvek® 400 D garments combine the protection, durability and comfort of DuPont™ Tyvek® fabric on the front and the comfort, softness and breathability of DuPont™ ProShield® 10 fabric on the back.
  • Comfort fit design
  • Laydown collar
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
  • White front, blue back
The DuPont™ Tyvek® 400 D garments are suitable for applications where comfort requirements are combined with limited protective requirements for frontal exposures. Garments are available in a variety of styles. Applications include fiberglassMore Product Information