DuPont™ ProShield® 10 Coveralls with Standard Hood
Supplier: DuPont
PB127SBU2X002500 PB127SBU3X002500 PB127SBU4X002500 PB127SBU5X002500 PB127SBU6X002500 PB127SBU7X002500 PB127SBULG002500 PB127SBUMD002500 PB127SBUSM002500 PB127SBUXL002500 PB127SWH2X002500 PB127SWH3X002500 PB127SWH4X002500 PB127SWH5X002500 PB127SWH6X002500 PB127SWH7X002500 PB127SWHLG002500 PB127SWHMD002500 PB127SWHSM002500 PB127SWHXL002500
10016-262CS 157.47 CAD
10016-262 10016-264 10016-266 10016-268 10016-270 10016-272 10016-274 10016-748 10016-750 10016-752 10016-754 10016-756 10016-758 10016-760 10016-762 10016-764 10016-766 10016-768 10016-770 10016-772
DuPont™ ProShield® 10 Coveralls with Standard Hood
Disposable Garments
DuPont™ ProShield® 10, spun bond melt blown spun bond (SMS) garments, are made to the high-quality standards you have come to expect from DuPont.
  • Standard fit hood
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments.
DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a varietyMore Product Information