DuPont™ ProShield® 70 Boot Covers with Skid Resistant Sole
Supplier: DuPont
P3452SGY00010000
10016-146CS 342.76 CAD
10016-146
DuPont™ ProShield® 70 Boot Covers with Skid Resistant Sole
Disposable Garments
ProShield® 70 fabric, used exclusively for shoe and boot cover products, is a spunbonded polypropylene substrate that is coated with a low-density polyethylene film for enhanced skid resistance and barrier performance.
  • Skid-resistant sole
  • Coverage up to calf
  • Serged seams
  • One size fits most
These boot covers feature a ProShield® 70 skid-resistant sole for enhanced skid resistance and coverage up to the calf.