Extraction Cell Bodies for ASE® Systems, Restek
Supplier: Restek
26132 26176 26178 25993 25994 25995 25996 26098 26110 26112 26114 26116
10015-262EA 277.11 CAD
10015-262 10015-280 10015-282 10015-384 10015-386 10015-388 10015-390 10015-420 10015-424 10015-426 10015-428 10015-430
Extraction Cell Bodies for ASE® Systems, Restek
Chromatography Supplies Chromatography Spare Parts
Accelerated solvent extraction is a common technique for fast and reliable extraction of organic materials from solid matrices, using EPA SW-846 Method 3545, Pressurized Fluid Extraction (PFE).
  • Cell bodies are serialized for easy sample identification
  • Smooth inner surfaces for easier cleaning
Restek offers a wide range of replacement parts to keep your extraction system running smoothly. All parts are economically priced to save you money and are designed to meet or exceed the performance of the original manufacturer’s parts.
Order Now
SPECIFICATIONS
Environmentally Preferable