ScanLater™ Western Blot Assay Kit, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
R8202 R8203 R8204 R8208 R8209 R8212 R8205 R8207 R8206 R8200 R8201 R8202 R8203 R8204 R8208 R8209 R8212 R8205 R8207 R8206 R8200 R8201 R8226 R8220 R8226 R8220
10048-854EA 587.25 CAD
10048-854 10048-856 10048-858 10048-860 10048-862 10048-864 10048-868 10048-870 10048-872 10048-936 10048-938 CA10048-854 CA10048-856 CA10048-858 CA10048-860 CA10048-862 CA10048-864 CA10048-868 CA10048-870 CA10048-872 CA10048-936 CA10048-938 10014-884 10014-888 CA10014-884 CA10014-888
ScanLater™ Western Blot Assay Kit, Molecular Devices
Blotting Kits
The ScanLater™ Western Blot Assay Kit is a time-resolved fluorescence (TRF)-based assay optimal for quantitating as little as femtogram protein samples.
  • Time-resolved fluorescence detection
  • Supports nitrocellulose and PVDF membranes
  • 340/80 nm excitation with 616/10 nm emission range
  • Optimized for use with ScanLater™
Using this novel western blot method, researchers can eliminate time-dependent substrate addition steps and sustain blot signal stability for at least one month
Europium-labeled goat, anti-rabbit, anti-mouse or streptavidin labeledMore Product Information