Axygen® Hydrophobic Filter Barrier Pipette Tips, Corning
Supplier: Corning
TF-1000-R-S TF-100-R-S TF-150-R-S TF-200-R-S TF-20-R-S TF-300-R-S TF-400-R-S TF-50-R-S MTXF-1250-C-R-S TXLF-10 TXLF-10-R-S TF-420-R-S TF-205-WB-R-S TF-1005-WB-R-S
10011-350CS 1216.19 CAD
10011-350 10011-352 10011-354 10011-356 10011-358 10011-360 10011-364 10011-370 12000-748 47745-490 47745-492 47749-872 80082-898 80082-900
Axygen® Hydrophobic Filter Barrier Pipette Tips, Corning
Pipette Tips
Filtered tips are ideal for molecular and microbiology applications in which carryover contamination can invalidate results.
  • Tips are metal-free
  • Filters are made from polyethylene and contain no PCR inhibitors
Tips are designed to fit a wide variety of single-channel and multichannel pipettors. Flexible walls and a series of internal sealing rings ensure a secure fit and require less force to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
All tips, except ExtendedMore Product Information