Axygen® Research-Grade Pipette Tips, Corning
Supplier: Corning
T-1000-B T-1000-B-R T-1000-B-R-S T-1005-WB-C-R T-1005-WB-C-R-S T-200-C-R T-200-C-R-S T-200-Y-R T-200-Y-R-S T-205-WB-C-R T-205-WB-C-R-S T-300-R T-300-R-S T-350-C-R T-400-R TR-222-C-R TR-222-C-R-S T-300-STK T-350-C T-200-C T-200-Y T-205-WB-C T-300 T-400 TR-222-C MTX-1250-C MTX-1250-C-R T-300-STK-S TXL-10 TXL-10-R TXL-10-R-S
10011-300CS 510.78 CAD
10011-300 10011-302 10011-304 10011-312 10011-314 10011-316 10011-318 10011-320 10011-322 10011-324 10011-326 10011-336 10011-338 10011-340 10011-344 10011-372 10011-374 10011-874 10011-876 10011-918 10011-920 10011-922 10011-928 10011-930 10011-932 12000-742 12000-744 47734-724 47745-484 47745-486 47745-488
Axygen® Research-Grade Pipette Tips, Corning
Pipette Tips
Tips are designed to fit a wide variety of single-channel and multichannel pipettors.
  • RNase-, DNase-, and endotoxin-free
Presterilized tips are exposed to e-beam radiation for protection from contamination.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Ordering information: Graduated tips have graduations at 10, 50, and 100 µl. The 200 µl wide-bore tips have a 1.More Product Information