Glassware Washers Accessories
Supplier: SP Industries
446508 10002-904 10002-906 10002-908 10002-910 10002-912 10002-914 10002-916 10002-918 10002-920 10002-922 10002-924 10002-926 10002-928 10002-930 10002-932 10002-934 10002-936 10002-938 10002-940 10002-942 10002-944
82021-092EA 2207.78 CAD
82021-092 10002-904 10002-906 10002-908 10002-910 10002-912 10002-914 10002-916 10002-918 10002-920 10002-922 10002-924 10002-926 10002-928 10002-930 10002-932 10002-934 10002-936 10002-938 10002-940 10002-942 10002-944
Glassware Washers Accessories
Glassware Washers
Accessories designed for use with VWR® Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
  • Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
  • Bottom mesh inserts prevent small items from falling through
  • Made of stainless steel for long life and durability
  • Booster pump for use with low pressure DI water supply below 18 psi
See details in table below for specific use information. For further assistance, view theMore Product Information