U-2D™ Micro-sample management system with racked inserts
Supplier: MicroSolv Technology Corp
95296-10P-CS 95296-10P-EA 95296-30P-CS 95296-30P-EA 95296-350-1DP 95296-350-1P 95296-350-3DP 95296-350-3P 95296-350-CS 95296-350-D-CS 95296-350-D-PK 95296-350-PK 95296-SB-CS 95296-SB-PK 95296-U2D 95296-05N 95296-1ML-1P 95296-1ML-1R 95296-1ML-3P 95296-1ML-3R 95296-500-1P 95296-500-1R 95296-500-3P 95296-500-3R 95296-500-CS 95296-1ML-PK 95296-500-PK 95296-350-1R 95296-350-3R 95296-U2D-1ML 95296-U2D-500 95296-I-1ML 95296-I-500
10002-764CS 184.23 CAD
10002-764 10002-790 10002-792 10002-794 10002-796 10002-798 10002-800 10002-802 10002-804 10002-806 10002-808 10002-810 10002-824 10002-826 10014-946 75854-082 76040-444 76040-448 76040-450 76040-452 76040-454 76040-456 76040-458 76040-460 76040-470 76040-476 76040-534 76045-326 76045-330 76045-348 76045-350 76057-650 76057-654
U-2D™ Micro-sample management system with racked inserts
Microplates Insert Plates and Inserts
Manage multiple low volume samples in a 96 well format and a unique split rack with glass inserts for vials.
  • 350 µl inserts can be used individually by dropping them into a 2 ml vial
  • Compatible with plate readers
  • Fill the inserts with multi-channel pipettors for quick and easy sample preparation
  • Transport inserts safely
Use this unique sample management system that is designed for preparing samples or for injecting them with autosamplers. Each rack is supplied with 96 glass inserts that are installed and ready for use. The included insert rack can beMore Product Information