Circulation Tubing Kit, Labconco®
Supplier: Labconco
5441808
10002-756EA 1113.9 CAD
10002-756
Circulation Tubing Kit, Labconco®
Tubing Transfer Tubing
Two each sections of 1.9 cm (³/₄") int.Ø flexible neoprene tubing with 1.9 cm (³/₄") ext.Ø brass compression nut tube fittings on each end.
Tubing kit is desgined for connecting Glove box monitor to Labconco® Glove box and AtmosPure™ Re-Gen gas purifier.