Introducing the Acrodisc One!
Give Your Samples the VIP Treatment from Pall Laboratory
Pall Acrodisc Premium Syringe Filters give you the certainty and assurance you need to ensure trouble-free analysis whatever your sample.
Acrodisc One™ is Pall’s key product for analytical sample preparation. The wwPTFE membrane in the Acrodisc One is made of hydrophilic PTFE and is the next generation of syringe filters for aqueous and aggressive organic solvent-based solutions.
Acrodisc PSF syringe filters are available with a GxF multi-layer prefilter which provides two to four times the throughput of standard glass fiber prefilter devices, allowing quick and easy filtration of your most difficult to filter solutions.
|