Masterflex® B/T® MasterSense™ BioPharma Process Pump
Reimage bioprocessing
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrowy
Shop nowConnected, efficient and durable
1. Built for high-flow, continuous duty, perfect for sanitary environments— 316 polished stainless steel housing
IP66 rated for washdown
2. Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
3. Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity - even with wet gloves on!
4. Over-the-air firmware updates ensure optimization of your pump system without moving the system while MasterflexLive® cloud storage system securely stores, monitors and generates reports for all Masterflex pump systems
5. Variable flow: 0.65 – 42 LPM
VIEW FULL SPECS
6. Qualitative tube calibration ensures tubing is calibrated precisely – choose PerfectPosition® pump tubiung sizes B/T 87 and B/T 91
SHOP B/T PUMP TUBING
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
Intuitive Operation
Simple touch screen interface, even while wearing gloves
- Easy to set up and program
- 5" capacitive TFT/RGB touch screen with a clear, wide viewing angle
- Adjust parameters with a simple touch—even with gloves on
- Operational modes include continuous run, volume dispense, and time dispense
- Batch control and interval/off time in Volume and Time modes
- Easily save and recall up to 50 commonly used protocols, each with a unique name
- Know when you hit your mark—graphical indicator lets you know when loaded tubing is optimally calibrated
- IP66 rated for washdown
Continuous Mode
Time Mode
Volume Mode
Remote Mode
Tube Calibration
User Access
Advanced Connectivity and Control Options
Remote, secure, and in compliance
Remote Monitoring
- MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
- Monitor and adjust critical processes running 24/7 without the need for onsite personnel
- Push notifications alert you to changes in operating conditions and error messages
Flexible Connectivity
- Your choice of connectivity options—MasterflexLive, EtherNet/IP, or Profibus
- Three-tier access to pump functionality—use this option to protect your process by controlling who can operate the pump and at what level
- Over-the-air firmware updates—automatically allows you to take full advantage of future enhancements
- Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
The Perfect Pump for High-Volume Applications
IP66 rated- built for sanitary, washdown environments including clean rooms, food, and dairy processing.
Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare.
Flows up to 42 LPM to ensure precise flow rates making this pump ideal for large-scale production.
Three-tier access and the ability to save and recall programs making your fluidic process secure and accurate.
![]() |
Item # MFLX77112-10 | |||
![]() ![]() ![]() ![]() ![]() What's Included Rapid-Load® pump head, 3 ft (1 m) of Tygon® LFL B/T 91 tubing 06430-91; and a 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. Unit is shipped with country specific plug/cord set; please specify ultimate destination when ordering. |
||||
Performance Specifications |
||||
Flow capacity | 0.65 to 42 LPM (0.17 to 11.1 GPM) | |||
rpm | 11 to 321 | |||
Number of heads accepted | 1 | |||
Maximum torque | Starting: 1130 N•cm (1600 oz-in) | |||
Running: 1017 N•cm (1440 oz-in) | ||||
Reversible | Yes | |||
External control - input | 0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Start/Stop, Direction (CW/CCW), Prime via contact closure | |||
External control - output | 0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm | |||
Network communication | EtherNet/IP, Profibus, MasterflexLive | |||
Electrical Specifications |
||||
Voltage | 90 to 260, universal voltage autoselecting | |||
Current | 5.0 A at 115 V; 3.1 A at 230 V | |||
Fuse rating | 6 A / 250 V | |||
Motor type | Continuous duty, TENV, BLDC | |||
Motor size | 370 W (0.5 hp) | |||
Display | 5” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels | |||
Motor/speed control type | PWM with microprocessor | |||
Speed regulation (repeatability) | ±0.1% | |||
Soft start/Electronic brake | Yes/Yes | |||
Physical specifications |
||||
Housing materials | 316 stainless steel housing, stainless steel chassis, stainless steel bezel | |||
Pump head materials | Powder-coat carbon steel and cast aluminum, stainless steel, polycarbonate, Nylatron® rollers | |||
IP rating | IP66 | |||
Agency listings | UL, cUL, CE, DLS | |||
Operating temperature | 0 to 40°C (32 to 104°F) | |||
Storage temperature | -25 to 65°C (-13 to 149°F) | |||
Noise level | <75 dBA @ 1 m (39") | |||
Dimensions (L x W x H) | 62.2 x 40.1 x 47.8 cm (24.5" x 15.8" x 18.8") | |||
Shipping weight | 54.5 kg (120 lb) |