Original and Accurate Hydranal™ Karl Fischer Reagents
Leading the way in innovation and safety since 1979, Hydranal™ reagents for Karl Fischer (KF) titration have been recognized as the most trusted brand in the industry.
The Hydranal portfolio is made up of methanol and ethanol-based KF reagents free from pyridine. Reagents for volumetric and coulometric application are completed by a wide range of standards and auxiliaries.
Hydranal products are known for their high quality and consistency, and are backed by dual accreditation, ISO/IEC 17025 and ISO 17034.
More than 700 Hydranal lab reports can be downloaded from www.lab-honeywell.com/hydranal-search/