Free ELPRO Data Logger Contest!
Are you interested in trying a new, electronic method for temperature monitoring? Fill out the questionnaire below for a chance to receive a free LIBERO Ti1 Data Logger from ELPRO. Limited quantities available.
The LIBERO Ti1 USB Data Logger is a safe, easy, and inexpensive way to monitor and record a wide range of temperature conditions. This compliant and economic solution for continuous monitoring is ideal for many applications in areas such as lab freezers, refrigerators, temperature-only incubators, ambient room conditions, and/or deli cases. This Data Logger monitors 24/7, and automatically generates a PDF report without the need to purchase software or hardware. The LIBERO complies with the ISO standard 19005-1 document management for the long-term preservation of electronic documents; is 21 CFR Part 11 and WHO compliant.