All You Need for Genomics
All You Need for Genomics
It’s an exciting time to be working in the field of genomics with the advancement of gene therapies, personalized medicine and with technologies such as qPCR, next generation sequencing (NGS), and CRISPR/Cas9 genome editing but also a very busy one.
VWR strives to enable your research by gathering technical information, product information, and quick links to genomics related applications into one, comprehensive location. Additionally, VWR has developed an All You Need for Genomics catalog to help you quickly locate products. Whether you are looking for DNA or RNA isolation kits, cloning kits and vectors, competent cells, restriction and modifying enzymes, or next gen library kits, we have all you need so you can stay focused on what is really important to you – your work
Featured Products
Make Your Research Count Down to the Very Last Drop
Minimize your sample and maximimize your performance with METTLER TOLEDO solutions for your Lab:
- UV-Vis: The UV5Nano Spectrophotometer only requires 1 μL of sample for reliable measurements, automatically measures at two precisely defined pathlengths allowing a wide concentration range to be measured without further dilutions.
- pH: The SevenExcellence™ multi-channel meter allows measurements of 1, 2 or 3 parameters simultaneously, and provides unmatched pH performance, simple operation, and compliance.
- Microbalances: Excellence Micro and Ultra Microbalances display unsurpassed accuracy even with the smallest sample sizes.