Anti-CHD4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A25773-100UG A25773-50UG
77740-577EA 521.4 CAD
77740-577 77857-793
Anti-CHD4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: CHD4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol CHD4
Environmentally Preferable
Antigen synonyms ATP dependent helicase ATP-dependent helicase CHD 4|CHD-4|CHD4_HUMAN|Chromodomain helicase DNA binding protein 4|Chromodomain-helicase-DNA-binding protein 4|Mi 2 autoantigen 218 kDa protein|Mi 2b|Mi-2 autoantigen 218 kDa protein|Mi2 beta|Mi2-beta
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q14839
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Chromodomain-helicase-DNA-binding protein 4 protein (1-219AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR