Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A69498-100UG A69498-50UG
77840-719EA 521.4 CAD
77840-719 77840-720
Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: DCLRE1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR