Anti-BHLHE41 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A70098-100UG A70098-50UG
77841-860EA 521.4 CAD
77841-860 77841-861
Anti-BHLHE41 Rabbit Recombinant Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000

Type:
Antigen: BHLHE41
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR