2-Phenylcyclohexanone ≥98.0%

Supplier: TCI America

P1311-5G P1311-1G
TCP1311-005GEA 274.02 CAD
TCP1311-005G TCP1311-1G
2-Phenylcyclohexanone ≥98.0%
2-Phenylcyclohexanone

CAS Number: 1444-65-1
MDL Number: MFCD00001629
Molecular Formula: C12H14O
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 61
Flash Point (°C): 110

Formula: C₁₂H₁₄O
Boiling Pt: 90 °C (0.5 mmHg)
Melting Pt: 61 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001629
CAS Number: 1444-65-1
EINECS: 215-888-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74cb164cb5b6ffac50240a579af2da66f2e06ecd&vid=74cb164cb5b6ffac50240a579af2da66f2e06ecd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR