1-Heptanesulfonic acid sodium salt ≥98.0%

Supplier: TCI America

Synonyms: Sodium heptane-1-sulfonate, Sodium 1-heptanesulfonate, Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate

H0454-500G H0454-25G H0454-100G
TCH0454-500GEA 1994.62 CAD
TCH0454-500G TCH0454-025G TCH0454-100G
1-Heptanesulfonic acid sodium salt ≥98.0%
1-Heptanesulphonic acid sodium salt

CAS Number: 22767-50-6
MDL Number: MFCD00007543
Molecular Formula: C7H16O3S
Molecular Weight: 202.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300


Other Product Names / Synonyms: 1-Heptanesulfonic Acid Sodium Salt, Heptylsulfonic Acid Sodium Salt

Formula: H₃C(CH₂)₅CH₂SO₃Na
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5db19c10ab1daab0940617346d980c8191b7cbc&vid=e5db19c10ab1daab0940617346d980c8191b7cbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR