1,4-Dioxane ≥99.0% stabilized

Supplier: TCI America

Synonyms: Dioxanep-DioxaneGlycolethylether1,4-Diethylene dioxide1,4-Dioxacyclohexane

Stabilization: Stabilised with Stabilised

D0860-500G D0860-25G
TCD0860-500GEA 55.76 CAD
TCD0860-500G TCD0860-25G
1,4-Dioxane ≥99.0% stabilized
1,4-Dioxane

(stabilized with BHT)
CAS Number: 123-91-1
MDL Number: MFCD00006571
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 12
Freezing point (°C): 11
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1,4-Diethylene Dioxide

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101 °C
Melting Pt: 11.8 °C
Density: 1.04 (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00006571
CAS Number: 123-91-1
UN: 1165
ADR: 3,II
Merck Index: 13,03330
https://ca.vwr.com/stibo/web/std.lang.all/59/61/28975961.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR