2'-Aminoacetophenone

Supplier: TCI America
Warning

Synonyms: 1-(2-Aminophenyl)ethanone, o-Aminoacetophenone

A0250-25G A0250-5G
TCA0250-025GEA 102.7 CAD
TCA0250-025G TCA0250-5G
2'-Aminoacetophenone
2'-Aminoacetophenone

CAS Number: 551-93-9
MDL Number: MFCD00007717
Molecular Weight: 135.17
Form: Clear Liquid
Boiling point (°C): 252
Flash Point (°C): 110
Specific Gravity (20/20): 1.12

Formula: C₈H₉NO
Boiling Pt: 252 °C
Melting Pt: 20 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007717
CAS Number: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94e1f30b9d010c2a44b1e2153cf300430b44e2c0&vid=94e1f30b9d010c2a44b1e2153cf300430b44e2c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR