Dihydrogen dinitrosulfatoplatinate(II) solution (4 - 6% Pt)

Supplier: Thermo Scientific Chemicals
Danger

12627-NA 12627-NC 12627-NJ
CAAA12627-NAEA 575.12 CAD
CAAA12627-NA CAAA12627-NC CAAA12627-NJ
Dihydrogen dinitrosulfatoplatinate(II) solution (4 - 6% Pt)
Dihydrogen dinitrosulfatoplatinate(II)

Pt DNS Solution


CAS No.: 12033-81-7 Molecular Formula: H2Pt(NO2)2SO4 Formula Weight: 384.17(anhy) Physical Form: Liquid UN No.: 3264 MDL No.: MFCD00064668

Formula: H₂Pt(NO₂)₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064668
CAS Number: 12033-81-7
UN: 3264
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR