4-Aminocinnamic acid ≥98.0%
Supplier: TCI America
Synonyms:
3-(4-Aminophenyl)acrylic acid, 4-Aminocinnamic Acid, 3-(4-Aminophenyl)acrylic Acid
A0691-5G
A0691-25G
TCA0691-005GEA
205.4
CAD
TCA0691-005G
TCA0691-025G
4-Aminocinnamic acid ≥98.0%
4-Aminocinnamic acid
CAS Number: 2393-18-2
MDL Number: MFCD00017117
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 170
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(4-Aminophenyl)acrylic Acid
Formula:
C₉H₉NO₂ Melting Pt: 170 °C |
MDL Number:
MFCD00017117 CAS Number: 2393-18-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...