Chlorine (Free and Total) Color Disc Test Kit, Hach
Supplier: Hach
223101
CA2231-01EA 228.44 CAD
CA2231-01
Chlorine (Free and Total) Color Disc Test Kit, Hach
Environmental Test Kits
This color disc test kit uses powder DPD reagent that reacts with chlorine more quickly than tablet-form DPD, giving more accurate results. Powder DPD also has a considerable advantage over orthotolidine, a hazardous substance sometimes used as a free and total chlorine test reagent. Hach Chlorine Test Kits are manufactured by Hach to ensure compliance with stringent quality control standards. Kit contains 100 tests. Range: 0.1-3.5 Mg/L.
Quickly and accurately measures free and total chlorine levels in the field or in the lab.
Simple and fast color disc method.
Certifications: UL Listed.
Accessories information: Accessory kit includes plug and case.