L(+)-Potassium sodium tartrate tetrahydrate ≥99%, granular USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrateRochelle saltL(+)-Tartaric acid potassium sodium salt tetrahydratePotassium sodium tartrate tetrahydratePotassium sodium L(+)-tartrate tetrahydrate(+)-Potassium sodium L-tartrate tetrahydrateSodium potassium tartratePotassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate(R,R)-(+)-Potassium sodium tartrate tetrahydrate

PO201-50KGBL PO201-2.5KG PO201-500GM PO201-12KG
95035-206EA 348.58 CAD
95035-204 89078-968 95035-206 700006-138
L(+)-Potassium sodium tartrate tetrahydrate ≥99%, granular USP
L(+)-Potassium sodium tartrate tetrahydrate

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Density: ∼1.767 g/cm³ (20 °C)
MDL Number: MFCD00150989
CAS Number: 6381-59-5
Merck Index: 13,07755
https://ca.vwr.com/stibo/web/std.lang.all/98/18/2299818.jpg

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR