Gelatin
Supplier: Spectrum
GE105-500GM GE105-2.5KG GE105-45KGBL GE105-12KG GE105-12KG
CA700011-238EA 5578.65 CAD
95033-350 CA700011-238 95033-352 700011-238 700011-240
Gelatin
Proteins and Peptides
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Gelatin, 250 Bloom, Powder, NF is used pharmaceutical formulations as thickening agent and emulsifier. All spectrum chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.