Potassium bromide, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, polished both sides (drilled)
Supplier: Thermo Scientific Chemicals
38475-KT
38475-KT
CAAA38475-KTEA
169.06
CAD
AA38475-KT
CAAA38475-KT
Potassium bromide, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, polished both sides (drilled)
Potassium bromide
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 °C (1013 hPa) Melting Pt: 730 °C Density: 2.75 g/cm³ (20 °C) |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 Merck Index: 13,07701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...