Epredia™ Specimen Transport System-STS™ Prefilled and Empty Containers, Epredia
Supplier: Epredia
531802 599602 531801 534801 56201 56401 56601 56901 561201 59201 59401 59601 59901 591201 591801ZF 594801ZF 59201ZF 59401ZF 59601ZF 59901ZF 591201ZF 59202 59402 59902 59602 591202 531201 4090 4091 531202 53151 53152 53301 53302 5332 5333 534802 5357 53601 5364 53901 53902 591801 594801 599601-9
89404-864CS 499.93 CAD
89404-864 89404-866 CA41810-040 CA48218-780 CA48218-782 CA48218-784 CA48218-786 CA48218-788 CA48218-790 CA48218-792 CA48218-794 CA48218-796 CA48218-798 CA48218-800 CA48218-802 CA48218-804 CA48218-806 CA48218-810 CA48218-812 CA48218-814 CA48218-816 CA48218-818 CA48218-820 CA48218-822 CA48218-824 CA48218-826 CA48218-828 CA72550-080 CA83009-838 CA83009-840 CA83009-842 CA83009-844 CA83009-846 CA83009-854 CA83009-856 CA83009-858 CA83009-860 CA83009-862 CA83009-864 CA83009-866 CA83009-868 CA83009-870 CA83009-872 CA83009-874 CA83009-984 CA83009-986 CA83009-990 76183-200
Epredia™ Specimen Transport System-STS™ Prefilled and Empty Containers, Epredia
Sample Containers General Purpose Containers
Transport, store and retrieve pathology specimens in a safe and cost-effective manner.
  • Containers are available prefilled with the highest quality 10% neutral buffered formalin (NBF), zinc formalin, or formalin-alcohol (20%)
  • Containers feature a highly visible bright yellow label for patient information
  • Each container is marked with a lot code and expiration date
  • Bio-Tite® closure systems are dual-threaded to minimize leakage, making it virtually impossible to cross-thread
  • Containers are nonsterile and comply with all updated federal and state OSHA requirement
Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Epredia™ Specimen Transportation System (STS™).
Three unique packaging configurations allow for easy distribution and transport. The 53 Series Containers are packaged in easy to transport trays ideal for operating rooms and hospitals. These clear polystyrene containers have screw-top lids, and comply with all updated federal and state OSHA requirements. Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks. The 56 Series Containers feature the Bio-Tite™ closure system, which is dual-threaded, making it virtually impossible to cross-thread while minimizing leakage. Packaged in easy to transport trays, these comply with all updated federal and state OSHA requirements. They are labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks. The 59 Series Containers also feature Bio-Tite™ closure systems but are packaged in stack pack format to reduce excess packaging. They comply with all updated federal and state OSHA requirements and are labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks.
This system includes containers pre-filled with the highest quality 10% Neutral Buffered Formalin (NBF) and Zinc Formalin.
Ordering information: The polystyrene and polypropylene containers have screw-top lids; the polyethylene containers have snap-top lids. Prefilled containers are stack-packed or packaged in trays for easy distribution and transportation of containers; empty containers are bulk packed.