Ultrafiltration Membrane Disc Filters
Supplier: Cytiva
OM100043 OM010043 OM030062 OM050043 OM005076 OM100043 OM003076 OM030062 OM005043 OM001076 OM050076 OM010062 OM003043 OM003043 OM030076 OM010076 OM300043 OM005043 OM003076 OM050076 OM001043 OM300043 OM001076 OM005076 OM010043 OM100076 OM030043 OM001043 OM030043 OM050043 OM030076 OM100076
CA29600-152PK 815.26 CAD
CA29600-152 CA29600-154 29300-150 29300-152 29300-130 CA29300-160 29300-098 29300-154 CA29300-162 29300-132 CA29300-120 CA29300-100 29300-136 CA29300-122 29300-158 CA29300-166 CA29300-124 29300-138 CA29300-128 29300-160 29300-162 CA29300-150 CA29300-130 29300-120 CA29300-132 29300-100 29300-122 29300-166 29300-124 CA29300-136 CA29300-138 29300-128
Ultrafiltration Membrane Disc Filters
Filters Membrane Filters
Replacement membranes for conventional, nondisposable stirred cell systems.
  • For conventional systems
  • Variety of diameters available
  • Non sterile
Use for concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules. Also for bench or pilot separations prior to scale-up. Available in a broad range of membrane diameters to fit various sizes. Omega™ membrane discs provide unmatched high flow rates and recoveries, and can be washed and reused. Non sterile. May be sanitized using 70% ethanol or 200 ppm sodium hypochlorite.