Anti-shha Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62490-50UG A62490-100UG
77720-331EA 360.84 CAD
77720-331 77720-330
Anti-shha Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: shha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR