SP Wilmad-LabGlass Large Orifice Volumetric Pipettes, SP Industries
Supplier: SP Industries
LG-9355-122 LG-9355-126 LG-9355-104 LG-9355-102 LG-9355-120 LG-9355-110 LG-9355-136 LG-9355-132 LG-9355-124
14203-654EA 74.96 CAD
14203-654 14211-494 14202-270 14201-896 14211-248 14211-402 14212-416 14212-418 89003-668
SP Wilmad-LabGlass Large Orifice Volumetric Pipettes, SP Industries
Pipettes Graduated Pipettes
Calibrated 'to deliver' at 20 °C, these borosilicate glass, color coded pipettes have a large orifice and are fabricated to meet the requirements for accuracy, appearance, glass quality, and calibration line accuracy.
Certifications: Reference: Volumetric, Laboratory Grade, ASTM E-969