SP Bel-Art Petri Dish Rack, 100 mm, Clear TPX® Body, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18985-0100
47751-754EA 338.18 CAD
47751-754
SP Bel-Art Petri Dish Rack, 100 mm, Clear TPX® Body, Bel-Art Products, a part of SP
Racks Petri Dish Racks
Clear, TPX® rack with polypropylene posts securely holds up to fifty-four 100 mm Petri dishes.
  • Clear body
  • TPX® construction with polypropylene posts
  • Shipped flat and easy to assemble
Easily viewed through the clear base and divider, dishes can be removed with one hand.
Built-in handle provides easy transport, and rubber feet help prevent rack from sliding on the benchtop. Racks are shipped flat and can be assembled in minutes.