VWR® Repeating Dispensers
Supplier: VWR International
53481-009 53481-064 53481-097 53481-122 53481-155 53481-188 53481-213 53481-246 53481-280 53481-348 53481-370 53481-406 53481-461 53481-494 53481-520 71001-648
53481-009EA 1039.85 CAD
53481-009 53481-064 53481-097 53481-122 53481-155 53481-188 53481-213 53481-246 53481-280 53481-348 53481-370 53481-406 53481-461 53481-494 53481-520 71001-648
VWR® Repeating Dispensers
Bottle Top Tilt Measures
Dispensers are recommended for multiple test procedures as well as dispensing strong acids, alkalies, and volatile reagents.
  • Accurate to ±2%
  • Capable of rapid repeat transfers of fixed volumes
  • Easy filling and dispensation
The calibrated pipet heads automatically fill and deliver contents by simple pouring action for rapid repeat transfers of fixed volumes of solutions. To fill, the pipet head is tilted backwards. Excess liquid returns to the flask automatically. Tilting dispenser forward delivers the calibrated volume. Pipet heads ordered separately have [ST] 24/40 inner joint.
Ordering information: A complete unit consists of an interchangeable glass pipet head, Erlenmeyer flask with [ST] 24/40 ground joint, and poly joint clamp.