Walter Crucible Holder, PLASTICOID COMPANY INC
Supplier: Plasticoid Company
VCHWGS
24065-000PK 168.13 CAD
24065-000
Walter Crucible Holder, PLASTICOID COMPANY INC
Supports Support Sleeves
Rubber crucible holder for use with Gooch crucibles, or similar crucibles, in the neck of a standard suction flask.
  • For use with crucibles 20 to 35 mL, and flasks 250 to 1000 mL
Delivery information: Supplied with funnel.