Acid Burets, Auto Self-Zeroing, Glass Stopcock, AMK Glass
Supplier: AMK GLASS INC
TG-18280-02 TG-18280-03 TG-18280-04
17585-022EA 2363.92 CAD
17585-022 17585-044 17585-066
Acid Burets, Auto Self-Zeroing, Glass Stopcock, AMK Glass
Burettes
Borosilicate glass buret assembly for use with acids
The acid-resistant polypropylene adapter fits a standard 5-pint screw top reagent bottle.
Ordering information: Buret assembly includes: buret, rubber bulb, and bottle adapter. Bottle not included.