Methylene blue, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Basic Blue 9Swiss BlueMethylthioninium chlorideMethylene blue chloride3,7-Bis(dimethylamino)phenothiazin-5-ium chloride

457250-1GM 457250-1GM
CA80058-364EA 154.42 CAD
CA80058-364 80058-364
Methylene blue, Calbiochem®
Methylene blue

Inhibitor of soluble guanylate cyclase. Organic spin-trap agent which forms stable adducts with oxygen free radicals in solution; biological staining agent.

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180 °C
Density: 1.757 g/cm³ (22 °C)
MDL Number: MFCD00150008
CAS Number: 61-73-4
Merck Index: 12,06137

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR