Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PerCP)

Supplier: AAT BIOQUEST INC
100211T0 100211T1 100211T2
77023-324EA 282.17 CAD
77023-324 77023-150 77023-830
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PerCP)
Antibodies
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR